Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: six2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100275.1 | NP_001093745.1 | 1 - 2 | 818 | 75 | |
GenBank mRNA | BC135532.1 | - | 1 - 2 | 818 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |