Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101806.1 | NP_001095276.1 | 4 - 5 | 591 | 192 | |
GenBank mRNA | BC088543.1 | - | 4 - 5 | 582 | 192 | |
BC160502.1 | - | 4 - 5 | 643 | 192 | ||
CR942536.2 | - | 4 - 5 | 591 | 192 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |