Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101807.1 | NP_001095277.1 | 2 - 3 | 351 | 68 | |
GenBank mRNA | BC135134.1 | - | 2 - 3 | 351 | 68 | |
CT030100.3 | - | 2 - 3 | 334 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |