Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102684.1 | NP_001096154.1 | 3656 | 92 | ||
GenBank mRNA | BC129014.1 | - | 3656 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |