Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102686.1 | NP_001096156.1 | 7 - 8 | 2310 | 73 | |
XM_018091904.1 | XP_017947393.1 | 6 - 7 | 1373 | 73 | ||
GenBank mRNA | BC136039.1 | - | 7 - 8 | 2310 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |