Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102707.1 | NP_001096177.1 | 7 - 8 | 1075 | 100 | |
GenBank mRNA | BC136176.1 | - | 7 - 8 | 1075 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |