Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acbd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102728.1 | NP_001096198.1 | 2 - 3 | 391 | 82 | |
XM_012962605.2 | XP_012818059.1 | 2 - 3 | 497 | 82 | ||
GenBank mRNA | BC135953.1 | - | 2 - 3 | 391 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |