Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zc3h6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102732.2 | NP_001096202.2 | 11 - 12 | 1762 | 98 | |
GenBank mRNA | AL866229.2 | - | 2 - 3 | 258 | 98 | |
BC135562.1 | - | 11 - 12 | 1754 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |