Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102762.1 | NP_001096232.1 | 4 - 5 | 525 | 61 | |
GenBank mRNA | BC135340.1 | - | 4 - 5 | 525 | 61 | |
BC155857.1 | - | 4 - 5 | 503 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |