Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102829.1 | NP_001096299.1 | 2 - 3 | 282 | 96 | |
XM_012962617.2 | XP_012818071.1 | 2 - 3 | 511 | 96 | ||
XM_018093760.1 | XP_017949249.1 | 2 - 3 | 378 | 96 | ||
GenBank mRNA | BC135622.1 | - | 2 - 3 | 282 | 96 | |
CT025221.2 | - | 2 - 3 | 197 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |