Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnaseh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102829.1 | NP_001096299.1 | 3 - 4 | 412 | 92 | |
XM_012962617.2 | XP_012818071.1 | 3 - 4 | 641 | 92 | ||
XM_018093760.1 | XP_017949249.1 | 3 - 4 | 508 | 92 | ||
GenBank mRNA | BC135622.1 | - | 3 - 4 | 412 | 92 | |
CT025221.2 | - | 3 - 4 | 327 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |