Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102838.1 | NP_001096308.1 | 2 - 3 | 345 | 103 | |
GenBank mRNA | BC135661.1 | - | 2 - 3 | 345 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |