Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102845.1 | NP_001096315.1 | 601 | 102 | ||
GenBank mRNA | BC135687.1 | - | 601 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |