Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ldlrad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102860.1 | NP_001096330.1 | 2 - 3 | 663 | 61 | |
GenBank mRNA | BC064324.1 | - | 2 - 3 | 645 | 61 | |
BC135753.1 | - | 2 - 3 | 663 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |