Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nova1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102869.1 | NP_001096339.1 | 3 - 4 | 654 | 64 | |
XM_012967886.2 | XP_012823340.1 | 3 - 4 | 580 | 64 | ||
GenBank mRNA | BC135781.1 | - | 3 - 4 | 654 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |