Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lysmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102871.1 | NP_001096341.1 | 1 - 2 | 403 | 72 | |
XM_012969816.2 | XP_012825270.1 | 2 - 3 | 390 | 72 | ||
GenBank mRNA | BC135785.1 | - | 1 - 2 | 403 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |