Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc45a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102873.1 | NP_001096343.1 | 5 - 6 | 1541 | 76 | |
GenBank mRNA | BC135794.1 | - | 5 - 6 | 1541 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |