Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prtfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102892.1 | NP_001096362.1 | 9 - 10 | 676 | 100 | |
XM_018094511.1 | XP_017950000.1 | 9 - 10 | 957 | 100 | ||
XM_018094512.1 | XP_017950001.1 | 8 - 9 | 936 | 100 | ||
GenBank mRNA | BC135868.1 | - | 9 - 10 | 676 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |