Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC135929.1 | - | 4 - 5 | 1033 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |