Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtkn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102913.1 | NP_001096383.1 | 5 - 6 | 607 | 86 | |
XM_012965162.1 | XP_012820616.1 | 5 - 6 | 1197 | 86 | ||
XM_012965199.1 | XP_012820653.1 | 5 - 6 | 607 | 86 | ||
XM_012965220.2 | XP_012820674.1 | 5 - 6 | 572 | 86 | ||
GenBank mRNA | BC135966.1 | - | 5 - 6 | 607 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |