Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfml2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102975.1 | NP_001096445.1 | 7 - 8 | 1455 | 76 | |
GenBank mRNA | BC136146.1 | - | 7 - 8 | 1455 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |