Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kansl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103023.1 | NP_001096493.1 | 8 - 9 | 1410 | 96 | |
XM_012956313.2 | XP_012811767.1 | 9 - 10 | 96 | |||
XM_012956314.2 | XP_012811768.1 | 9 - 10 | 96 | |||
XM_012956315.2 | XP_012811769.1 | 9 - 10 | 96 | |||
XM_018091048.1 | XP_017946537.1 | 9 - 10 | 96 | |||
XM_018091049.1 | XP_017946538.1 | 9 - 10 | 96 | |||
GenBank mRNA | BC135559.1 | - | 8 - 9 | 1410 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |