Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wasf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103026.1 | NP_001096496.1 | 2 - 3 | 158 | 87 | |
XM_018091050.1 | XP_017946539.1 | 2 - 3 | 162 | 87 | ||
GenBank mRNA | BC135144.1 | - | 2 - 3 | 158 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |