Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103048.1 | NP_001096518.1 | 4 - 5 | 1406 | 81 | |
GenBank mRNA | BC135591.1 | - | 4 - 5 | 1406 | 81 | |
CT025512.2 | - | 4 - 5 | 1405 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |