Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mad2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103058.1 | NP_001096528.1 | 3 - 4 | 380 | 79 | |
GenBank mRNA | BC135782.1 | - | 3 - 4 | 380 | 79 | |
CT025283.2 | - | 3 - 4 | 336 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |