Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103062.1 | NP_001096532.1 | 7 - 8 | 924 | 87 | |
XM_012969992.2 | XP_012825446.1 | 7 - 8 | 87 | |||
XM_012969993.2 | XP_012825447.1 | 8 - 9 | 87 | |||
XM_018097025.1 | XP_017952514.1 | 8 - 9 | 87 | |||
XM_018097026.1 | XP_017952515.1 | 7 - 8 | 87 | |||
XM_018097027.1 | XP_017952516.1 | 7 - 8 | 87 | |||
GenBank mRNA | BC135858.1 | - | 7 - 8 | 924 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |