Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272027.1 | NP_001258956.1 | 8 - 9 | 1841 | 71 | |
XM_012963038.2 | XP_012818492.1 | 9 - 10 | 2057 | 71 | ||
XM_012963039.2 | XP_012818493.1 | 9 - 10 | 2114 | 71 | ||
XM_018093904.1 | XP_017949393.1 | 9 - 10 | 2084 | 71 | ||
GenBank mRNA | BC121341.1 | - | 4 - 5 | 632 | 71 | |
BC161191.1 | - | 8 - 9 | 1841 | 71 | ||
CR848654.2 | - | 3 - 4 | 582 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |