Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272027.1 | NP_001258956.1 | 10 - 11 | 2093 | 107 | |
XM_012963038.2 | XP_012818492.1 | 11 - 12 | 2309 | 107 | ||
XM_012963039.2 | XP_012818493.1 | 11 - 12 | 2366 | 107 | ||
XM_018093904.1 | XP_017949393.1 | 11 - 12 | 2336 | 107 | ||
GenBank mRNA | BC121341.1 | - | 6 - 7 | 884 | 107 | |
BC161191.1 | - | 10 - 11 | 2093 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |