Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110042.1 | NP_001103512.1 | 4 - 5 | 730 | 85 | |
XM_012954239.1 | XP_012809693.1 | 4 - 5 | 808 | 85 | ||
GenBank mRNA | BC153713.1 | - | 4 - 5 | 730 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |