Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec61b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110044.1 | NP_001103514.1 | 2 - 3 | 195 | 75 | |
GenBank mRNA | BC135617.2 | - | 2 - 3 | 195 | 75 | |
CR942631.1 | - | 2 - 3 | 131 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |