Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110045.1 | NP_001103515.1 | 3 - 4 | 747 | 75 | |
XM_018096095.1 | XP_017951584.1 | 3 - 4 | 839 | 75 | ||
GenBank mRNA | BC136082.1 | - | 3 - 4 | 741 | 75 | |
BC153333.1 | - | 3 - 4 | 747 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |