Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bmp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112907.1 | NP_001106378.1 | 6 - 7 | 1219 | 77 | |
GenBank mRNA | BC155509.1 | - | 6 - 7 | 1219 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |