Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: avd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112934.1 | NP_001106405.1 | 1 - 2 | 98 | 76 | |
GenBank mRNA | BC152704.1 | - | 1 - 2 | 98 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |