Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112940.2 | NP_001106411.2 | 3 - 4 | 449 | 97 | |
XM_012954147.2 | XP_012809601.1 | 3 - 4 | 548 | 97 | ||
XM_012954148.2 | XP_012809602.1 | 3 - 4 | 548 | 97 | ||
XM_018095238.1 | XP_017950727.1 | 3 - 4 | 547 | 97 | ||
XM_018095239.1 | XP_017950728.1 | 3 - 4 | 552 | 97 | ||
GenBank mRNA | BC153342.1 | - | 3 - 4 | 474 | 97 | |
BC161065.1 | - | 3 - 4 | 449 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |