Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jmy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112955.1 | NP_001106426.1 | 7 - 8 | 1591 | 81 | |
GenBank mRNA | BC153370.1 | - | 7 - 8 | 1591 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |