Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thumpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112958.1 | NP_001106429.1 | 9 - 10 | 1185 | 102 | |
XM_012962651.2 | XP_012818105.1 | 10 - 11 | 1317 | 102 | ||
XM_012962652.2 | XP_012818106.1 | 10 - 11 | 1254 | 102 | ||
GenBank mRNA | BC153707.1 | - | 9 - 10 | 1185 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |