Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam207a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112959.1 | NP_001106430.1 | 5 - 6 | 570 | 92 | |
XM_018097030.1 | XP_017952519.1 | 4 - 5 | 371 | 92 | ||
GenBank mRNA | BC153710.1 | - | 5 - 6 | 570 | 92 | |
BC158169.1 | - | 5 - 6 | 572 | 92 | ||
CR761930.2 | - | 5 - 6 | 569 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |