Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: naa60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112973.1 | NP_001106444.1 | 4 - 5 | 427 | 67 | |
XM_012969997.2 | XP_012825451.1 | 3 - 4 | 431 | 67 | ||
GenBank mRNA | BC153734.1 | - | 4 - 5 | 427 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |