Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tp53i11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112976.1 | NP_001106447.1 | 2 - 3 | 533 | 66 | |
GenBank mRNA | BC153738.1 | - | 2 - 3 | 533 | 66 | |
CU075620.1 | - | 2 - 3 | 327 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |