Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2u1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113000.1 | NP_001106471.1 | 4 - 5 | 1601 | 98 | |
GenBank mRNA | BC154092.1 | - | 4 - 5 | 1601 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |