Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: begain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113026.2 | NP_001106497.2 | 8 - 9 | 712 | 80 | |
XM_012967927.2 | XP_012823381.1 | 7 - 8 | 808 | 80 | ||
XM_012967928.2 | XP_012823382.1 | 6 - 7 | 718 | 80 | ||
XM_012967929.2 | XP_012823383.1 | 7 - 8 | 808 | 80 | ||
XM_018096129.1 | XP_017951618.1 | 4 - 5 | 769 | 80 | ||
GenBank mRNA | BC154696.1 | - | 8 - 9 | 712 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |