Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lypd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113032.3 | NP_001106503.1 | 1 - 2 | 194 | 96 | |
GenBank mRNA | BC154706.1 | - | 392 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |