Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmprss12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113037.1 | NP_001106508.1 | 3 - 4 | 381 | 62 | |
XM_012956358.2 | XP_012811812.1 | 2 - 3 | 926 | 62 | ||
GenBank mRNA | BC154712.1 | - | 3 - 4 | 381 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |