Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmigd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113059.1 | NP_001106530.1 | 2 - 3 | 163 | 109 | |
GenBank mRNA | BC154899.1 | - | 2 - 3 | 163 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |