Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c17orf67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113068.1 | NP_001106539.1 | 1 - 2 | 84 | 73 | |
GenBank mRNA | BC154910.1 | - | 1 - 2 | 84 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |