Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113081.1 | NP_001106552.1 | 3 - 4 | 356 | 76 | |
GenBank mRNA | BC154931.1 | - | 3 - 4 | 356 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |