Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tm7sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113126.1 | NP_001106597.1 | 7 - 8 | 906 | 64 | |
XM_012960551.2 | XP_012816005.1 | 10 - 11 | 1139 | 64 | ||
XM_012960552.2 | XP_012816006.1 | 8 - 9 | 1032 | 64 | ||
XM_012960553.2 | XP_012816007.1 | 9 - 10 | 1109 | 64 | ||
GenBank mRNA | BC155427.1 | - | 7 - 8 | 906 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |