Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113666.1 | NP_001107138.1 | 2 - 3 | 308 | 139 | |
GenBank mRNA | BC157161.1 | - | 2 - 3 | 308 | 139 | |
BC170646.1 | - | 2 - 3 | 300 | 139 | ||
BC170650.1 | - | 2 - 3 | 300 | 139 | ||
CT025345.2 | - | 2 - 3 | 134 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |