Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113671.1 | NP_001107143.1 | 1 - 2 | 233 | 106 | |
GenBank mRNA | AM041993.1 | - | 1 - 2 | 233 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Xenopus tropicalis Probe spans exons |