Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rerg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113676.1 | NP_001107148.1 | 6 - 7 | 625 | 77 | |
XM_012958519.2 | XP_012813973.1 | 3 - 4 | 469 | 77 | ||
GenBank mRNA | BC155994.1 | - | 6 - 7 | 625 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |